Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTCATTACAAAAATGTTGAACG[C/T]TAATTGTTTTTTTAAAATACAATTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603659 | ||||||||||||||||||||
Literature Links: |
GLP2R PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GLP2R - glucagon like peptide 2 receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004246.2 | Intron | NP_004237.1 | ||||
XM_005256861.3 | Intron | XP_005256918.1 | ||||
XM_011524077.2 | Intron | XP_011522379.1 | ||||
XM_017025339.1 | Intron | XP_016880828.1 | ||||
XM_017025340.1 | Intron | XP_016880829.1 | ||||
XM_017025341.1 | Intron | XP_016880830.1 |
GSG1L2 - GSG1 like 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |