Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGACTGTGTTGCACAGTCACTCC[A/G]TTTTTTGGTGCTGTCTTGTAGGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
19 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607983 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GORAB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GORAB - golgin, RAB6 interacting | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001146039.1 | Intron | NP_001139511.1 | ||||
NM_001320252.1 | Intron | NP_001307181.1 | ||||
NM_152281.2 | Intron | NP_689494.2 | ||||
XM_006711628.3 | Intron | XP_006711691.1 | ||||
XM_011510149.2 | Intron | XP_011508451.1 | ||||
XM_011510150.2 | Intron | XP_011508452.1 | ||||
XM_011510151.1 | Intron | XP_011508453.1 | ||||
XM_017002807.1 | Intron | XP_016858296.1 |
LOC101928650 - uncharacterized LOC101928650 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |