Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGATGCTTAGTTCAAAGCTGATA[C/T]ATTGGTTTGCTTCTGTGTCCCCACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 102642 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SOAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SOAT1 - sterol O-acyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001252511.1 | Intron | NP_001239440.1 | ||||
NM_001252512.1 | Intron | NP_001239441.1 | ||||
NM_003101.5 | Intron | NP_003092.4 | ||||
XM_011509911.1 | Intron | XP_011508213.1 | ||||
XM_011509912.1 | Intron | XP_011508214.1 |
Set Membership: |
HapMap |