Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCAACTGTCTCTGTCCGCGGTCC[C/T]GGGTCCCCGGTGGCGGCAAGAGAGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP350 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CEP350 - centrosomal protein 350 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014810.4 | Intron | NP_055625.4 | ||||
XM_011510191.2 | Intron | XP_011508493.1 | ||||
XM_011510198.2 | Intron | XP_011508500.1 | ||||
XM_017002929.1 | Intron | XP_016858418.1 | ||||
XM_017002930.1 | Intron | XP_016858419.1 | ||||
XM_017002931.1 | Intron | XP_016858420.1 |
LOC101928933 - uncharacterized LOC101928933 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |