Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGAGGATGAGGAGGTGCAACAGC[C/G]GCTCCGGGCCGCCGCCGTCGCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603120 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
QSOX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
QSOX1 - quiescin sulfhydryl oxidase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004128.2 | 94 | Missense Mutation | CGC,GGC | R,G 7 | NP_001004128.1 | |
NM_002826.4 | 94 | Missense Mutation | CGC,GGC | R,G 7 | NP_002817.2 |