Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGTGAAACCAGTGGGGCTGACC[A/G]TATATATATGTGTGTATACAAGTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
29 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600889 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFHR2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CFHR2 - complement factor H related 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312672.1 | Intron | NP_001299601.1 | ||||
NM_005666.3 | Intron | NP_005657.1 | ||||
XM_005245113.3 | Intron | XP_005245170.1 | ||||
XM_011509458.2 | Intron | XP_011507760.1 | ||||
XM_011509459.2 | Intron | XP_011507761.1 | ||||
XM_011509460.2 | Intron | XP_011507762.1 | ||||
XM_017001109.1 | Intron | XP_016856598.1 |