Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTTCATAGTTTGAGAATGTTTTT[C/T]CTAAATTTTCTTTGATCTGCTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151460 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRC - protein tyrosine phosphatase, receptor type C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267798.1 | Intron | NP_001254727.1 | ||||
NM_002838.4 | Intron | NP_002829.3 | ||||
NM_080921.3 | Intron | NP_563578.2 | ||||
XM_006711472.3 | Intron | XP_006711535.1 | ||||
XM_006711473.3 | Intron | XP_006711536.1 | ||||
XM_006711474.3 | Intron | XP_006711537.1 |