Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTTGTAACCATTTTGTGACAACT[A/G]CATATATATTATTCACAGAGATTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ATP6V1G3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP6V1G3 - ATPase H+ transporting V1 subunit G3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320218.1 | Intron | NP_001307147.1 | ||||
NM_133262.2 | Intron | NP_573569.1 | ||||
NM_133326.1 | Intron | NP_579872.1 | ||||
XM_006711163.3 | Intron | XP_006711226.1 | ||||
XM_011509186.2 | Intron | XP_011507488.1 | ||||
XM_011509187.2 | Intron | XP_011507489.1 |