Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGACCTGGATGTGCAGGATGGATCC[A/G]GGGAGGGTAGGGAAAAAGGATGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGFN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
IGFN1 - immunoglobulin-like and fibronectin type III domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164586.1 | Intron | NP_001158058.1 | ||||
XM_005245580.3 | Intron | XP_005245637.1 | ||||
XM_006711618.3 | Intron | XP_006711681.1 | ||||
XM_011510120.2 | Intron | XP_011508422.1 | ||||
XM_011510121.2 | Intron | XP_011508423.1 | ||||
XM_017002787.1 | Intron | XP_016858276.1 |