Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTTGAAGGACCAAAGGAAAAGAG[A/C]AAAAACCAGCCTCTAACCAGGCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 173445 | ||||||||||||||||||||
Literature Links: |
PECAM1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PECAM1 - platelet and endothelial cell adhesion molecule 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000442.4 | 3086 | UTR 3 | NP_000433.4 | |||
XM_005276880.1 | 3086 | UTR 3 | XP_005276937.1 | |||
XM_005276881.1 | 3086 | UTR 3 | XP_005276938.1 | |||
XM_005276882.1 | 3086 | UTR 3 | XP_005276939.1 | |||
XM_005276883.2 | 3086 | Intron | XP_005276940.1 | |||
XM_011524889.2 | 3086 | UTR 3 | XP_011523191.1 | |||
XM_011524890.1 | 3086 | UTR 3 | XP_011523192.1 | |||
XM_017024738.1 | 3086 | UTR 3 | XP_016880227.1 | |||
XM_017024739.1 | 3086 | UTR 3 | XP_016880228.1 | |||
XM_017024740.1 | 3086 | UTR 3 | XP_016880229.1 | |||
XM_017024741.1 | 3086 | UTR 3 | XP_016880230.1 |