Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608938 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KB1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RPS6KB1 - ribosomal protein S6 kinase B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272042.1 | Intron | NP_001258971.1 | ||||
NM_001272043.1 | Intron | NP_001258972.1 | ||||
NM_001272044.1 | Intron | NP_001258973.1 | ||||
NM_001272060.1 | Intron | NP_001258989.1 | ||||
NM_003161.3 | Intron | NP_003152.1 | ||||
XM_011525101.2 | Intron | XP_011523403.1 | ||||
XM_011525102.2 | Intron | XP_011523404.1 | ||||
XM_011525103.2 | Intron | XP_011523405.1 | ||||
XM_017024929.1 | Intron | XP_016880418.1 | ||||
XM_017024930.1 | Intron | XP_016880419.1 | ||||
XM_017024931.1 | Intron | XP_016880420.1 | ||||
XM_017024932.1 | Intron | XP_016880421.1 | ||||
XM_017024933.1 | Intron | XP_016880422.1 |
Set Membership: |
HapMap |