Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGAGCGGGACCGGGCTGTCACCGG[C/G]AGCGGCGGAAGGAAGGGGGTCACAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
18 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 116952 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDC42 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CDC42 - cell division cycle 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039802.1 | Intron | NP_001034891.1 | ||||
NM_001791.3 | Intron | NP_001782.1 | ||||
NM_044472.2 | Intron | NP_426359.1 |
Set Membership: |
HapMap |