Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAACAGAGGCAAATGAAAAAACA[A/C]TTCAAATTAATTCTACAAAGATTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606264 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLEC7A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
CLEC7A - C-type lectin domain family 7 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022570.4 | Intron | NP_072092.2 | ||||
NM_197947.2 | Intron | NP_922938.1 | ||||
NM_197948.2 | Intron | NP_922939.1 | ||||
NM_197949.2 | Intron | NP_922940.1 | ||||
NM_197950.2 | Intron | NP_922941.1 | ||||
NM_197954.2 | Intron | NP_922945.1 | ||||
XM_006719135.3 | Intron | XP_006719198.1 | ||||
XM_017019822.1 | Intron | XP_016875311.1 | ||||
XM_017019823.1 | Intron | XP_016875312.1 |