Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601210 | |||||||||||||||||||||||
Literature Links: |
PCBP2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PCBP2 - poly(rC) binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098620.2 | Intron | NP_001092090.1 | ||||
NM_001128911.1 | Intron | NP_001122383.1 | ||||
NM_001128912.1 | Intron | NP_001122384.1 | ||||
NM_001128913.1 | Intron | NP_001122385.1 | ||||
NM_001128914.1 | Intron | NP_001122386.1 | ||||
NM_005016.5 | Intron | NP_005007.2 | ||||
NM_031989.4 | Intron | NP_114366.1 |
PCBP2-OT1 - PCBP2 overlapping transcript 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |