Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATTTTTTAGTTAGGGGGAACA[A/G]AGTCTCATTCTGTCACCCAGGCTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
RNF41 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RNF41 - ring finger protein 41 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242826.1 | Intron | NP_001229755.1 | ||||
NM_005785.3 | Intron | NP_005776.1 | ||||
NM_194358.2 | Intron | NP_919339.1 | ||||
NM_194359.2 | Intron | NP_919340.1 | ||||
XM_005268561.4 | Intron | XP_005268618.1 | ||||
XM_011537734.2 | Intron | XP_011536036.1 | ||||
XM_011537735.2 | Intron | XP_011536037.1 |