Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGTACCTGTAAAAGGAGGTATGC[A/G]GTGCTTCTAAAGCATGCACTGCATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191175 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf73 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
C12orf73 - chromosome 12 open reading frame 73 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135570.1 | 1067 | UTR 3 | NP_001129042.1 | |||
XM_011538718.2 | 1067 | UTR 3 | XP_011537020.1 | |||
XM_017019916.1 | 1067 | UTR 3 | XP_016875405.1 | |||
XM_017019917.1 | 1067 | UTR 3 | XP_016875406.1 |
HSP90B1 - heat shock protein 90 beta family member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |