Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGAAACAACTAAAGAATGCCATAA[A/G]TCAACGTGGCACAAAAGGCCCCTAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611911 MIM: 611684 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISCU PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ISCU - iron-sulfur cluster assembly enzyme | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001301140.1 | Intron | NP_001288069.1 | ||||
NM_001301141.1 | Intron | NP_001288070.1 | ||||
NM_001320042.1 | Intron | NP_001306971.1 | ||||
NM_014301.4 | Intron | NP_055116.1 | ||||
NM_213595.3 | Intron | NP_998760.1 | ||||
XM_011538100.2 | Intron | XP_011536402.1 | ||||
XM_011538101.2 | Intron | XP_011536403.1 |
SART3 - squamous cell carcinoma antigen recognized by T-cells 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |