Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGTGAGGTAGCTTATCTGCTGGC[C/T]AGGAAGCCACGGAGGGCCTTCCCTA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D7 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SH2D7 - SH2 domain containing 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101404.1 | 1035 | Intron | NP_001094874.1 | |||
XM_011521895.2 | 1035 | UTR 5 | XP_011520197.1 | |||
XM_011521896.2 | 1035 | Intron | XP_011520198.1 | |||
XM_011521899.2 | 1035 | Intron | XP_011520201.1 | |||
XM_017022487.1 | 1035 | Intron | XP_016877976.1 | |||
XM_017022488.1 | 1035 | Intron | XP_016877977.1 |
Set Membership: |
HapMap |