Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGGGGCAGGGCAGAGAAGAGCAAG[C/G]CCATCCTCAGAGCAGAGGATGCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SH2D7 - SH2 domain containing 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D2B - TBC1 domain family member 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |