Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCACATTATATTTGTGTTTGTTCAG[A/T]TTTCTTATTAAGTGCAATAGTGTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
KIAA1586 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KIAA1586 - KIAA1586 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286274.1 | Intron | NP_001273203.1 | ||||
NM_001286275.1 | Intron | NP_001273204.1 | ||||
NM_001286276.1 | Intron | NP_001273205.1 | ||||
NM_020931.3 | Intron | NP_065982.1 | ||||
XM_011514767.2 | Intron | XP_011513069.1 |