Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAATCTCAGAAGCTTTTTAGCA[G/T]GAAGCCTATAAAGGATGAATATCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610525 | ||||||||||||||||||||
Literature Links: |
HPCAL4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HPCAL4 - hippocalcin like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282396.1 | 3399 | UTR 3 | NP_001269325.1 | |||
NM_001282397.1 | 3399 | UTR 3 | NP_001269326.1 | |||
NM_016257.3 | 3399 | UTR 3 | NP_057341.1 | |||
XM_017001447.1 | 3399 | UTR 3 | XP_016856936.1 |
NT5C1A - 5'-nucleotidase, cytosolic IA | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |