Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACGCCCCCGACGGCGAGCGCAGC[A/C]GCAGCCGCGCCCGCACCCGCAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 130500 | ||||||||||||||||||||
Literature Links: |
EPB41 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPB41 - erythrocyte membrane protein band 4.1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM200B - transmembrane protein 200B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001003682.3 | 214 | Missense Mutation | CGG,CTG | R,L 47 | NP_001003682.1 | |
NM_001171868.1 | 214 | Missense Mutation | CGG,CTG | R,L 47 | NP_001165339.1 | |
XM_011541457.2 | 214 | Missense Mutation | CGG,CTG | R,L 58 | XP_011539759.1 |