Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGATCTAGTGCTCCAGGTCGCT[C/T]GGAACCGCTTCAGTGCCTAGTTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608205 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MECR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MECR - mitochondrial trans-2-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024732.2 | Intron | NP_001019903.2 | ||||
NM_016011.3 | Intron | NP_057095.3 | ||||
XM_005245885.2 | Intron | XP_005245942.1 | ||||
XM_005245887.2 | Intron | XP_005245944.1 | ||||
XM_011541539.1 | Intron | XP_011539841.1 | ||||
XM_011541540.1 | Intron | XP_011539842.1 | ||||
XM_011541541.1 | Intron | XP_011539843.1 | ||||
XM_011541543.1 | Intron | XP_011539845.1 | ||||
XM_011541545.1 | Intron | XP_011539847.1 | ||||
XM_011541546.1 | Intron | XP_011539848.1 | ||||
XM_011541547.1 | Intron | XP_011539849.1 | ||||
XM_011541549.1 | Intron | XP_011539851.1 | ||||
XM_011541550.1 | Intron | XP_011539852.1 | ||||
XM_011541553.1 | Intron | XP_011539855.1 | ||||
XM_011541554.1 | Intron | XP_011539856.1 | ||||
XM_017001411.1 | Intron | XP_016856900.1 | ||||
XM_017001412.1 | Intron | XP_016856901.1 | ||||
XM_017001413.1 | Intron | XP_016856902.1 | ||||
XM_017001414.1 | Intron | XP_016856903.1 | ||||
XM_017001415.1 | Intron | XP_016856904.1 | ||||
XM_017001416.1 | Intron | XP_016856905.1 |
Set Membership: |
HapMap |