Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCAGTTCCGATGAAATGAGGTCAA[C/T]ATGACATGATCCTTTTGGAATGACT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604344 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAN1A1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAN1A1 - mannosidase alpha class 1A member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005907.3 | 2865 | UTR 3 | NP_005898.2 | |||
XM_005266986.4 | 2865 | UTR 3 | XP_005267043.1 | |||
XM_011535833.2 | 2865 | UTR 3 | XP_011534135.1 | |||
XM_011535834.2 | 2865 | Intron | XP_011534136.1 |
Set Membership: |
HapMap |