Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCCCCAGATTCTGTATCCTCCAT[A/G]GTCCTATTCATGACATGGGTCGTTC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603424 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101927027 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101927027 - uncharacterized LOC101927027 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRKRA - protein activator of interferon induced protein kinase EIF2AK2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139517.1 | Intron | NP_001132989.1 | ||||
NM_001139518.1 | Intron | NP_001132990.1 | ||||
NM_001316362.1 | Intron | NP_001303291.1 | ||||
NM_003690.4 | Intron | NP_003681.1 | ||||
XM_011512063.1 | Intron | XP_011510365.1 | ||||
XM_011512066.1 | Intron | XP_011510368.1 | ||||
XM_017005159.1 | Intron | XP_016860648.1 |