Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGTGAGCCACTGCGCCTGGTGATA[C/T]TGGATTTATATATCTGGAGTCTGAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610266 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371931 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC105371931 - uncharacterized LOC105371931 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4523 - microRNA 4523 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TAOK1 - TAO kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020791.2 | Intron | NP_065842.1 | ||||
NM_025142.1 | Intron | NP_079418.1 | ||||
XM_011525060.2 | Intron | XP_011523362.1 |
Set Membership: |
HapMap |