Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCGGGGCTGAGGCTGCGGGAGCT[G/A]GAGCGGGGAAGAAAAGGGAATTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 186591 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101928762 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LOC101928762 - uncharacterized LOC101928762 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STX4 - syntaxin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001272095.1 | 86 | UTR 5 | NP_001259024.1 | |||
NM_001272096.1 | 86 | Intron | NP_001259025.1 | |||
NM_004604.4 | 86 | UTR 5 | NP_004595.2 | |||
XM_005255521.1 | 86 | Intron | XP_005255578.1 | |||
XM_011545925.2 | 86 | Intron | XP_011544227.1 |
Set Membership: |
HapMap |