Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCAAAGGCTATTCAGAAGCAGA[A/G]ATTGAAAACTGGCAGCTCACTGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
71 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 134629 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FDPS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FDPS - farnesyl diphosphate synthase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135821.1 | Intron | NP_001129293.1 | ||||
NM_001135822.1 | Intron | NP_001129294.1 | ||||
NM_001242824.1 | Intron | NP_001229753.1 | ||||
NM_001242825.1 | Intron | NP_001229754.1 | ||||
NM_002004.3 | Intron | NP_001995.1 | ||||
XM_005244962.1 | Intron | XP_005245019.1 | ||||
XM_005244963.1 | Intron | XP_005245020.1 |
LOC105371451 - uncharacterized LOC105371451 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RUSC1 - RUN and SH3 domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RUSC1-AS1 - RUSC1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |