Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCGTTTCCTAATTACAGAGGCA[G/T]GATTGCACACATGCTCTAAACCACA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604539 | ||||||||||||||||||||
Literature Links: |
ADAMTS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ADAMTS2 - ADAM metallopeptidase with thrombospondin type 1 motif 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014244.4 | 5745 | UTR 3 | NP_055059.2 | |||
NM_021599.3 | 5745 | Intron | NP_067610.1 | |||
XM_017010063.1 | 5745 | UTR 3 | XP_016865552.1 |