Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGGATGACAAGAATGCAGCCTCAG[A/G]AAAAATGAACTGTATGAAATGTATC
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 608357 | |||||||||||||||||||||||||||||
Literature Links: |
SULT1C4 PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | JPT (Japanese)
|
|||
AFR - Not Available | Japanese - Not Available | CHB (Han Chinese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
SULT1C4 - sulfotransferase family 1C member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321770.1 | Intron | NP_001308699.1 | ||||
NM_006588.3 | Intron | NP_006579.2 | ||||
XM_017003807.1 | Intron | XP_016859296.1 |
Set Membership: |
HapMap |