Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATAATACATATACATGATTGTTA[C/T]ACATCTTTAGAAAGCAAAGAAAAAC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYLK4 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MYLK4 - myosin light chain kinase family member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005249078.3 | 5598 | UTR 3 | XP_005249135.2 | |||
XM_005249079.2 | 5598 | UTR 3 | XP_005249136.1 | |||
XM_006715082.3 | 5598 | UTR 3 | XP_006715145.1 | |||
XM_011514567.2 | 5598 | UTR 3 | XP_011512869.1 | |||
XM_017010819.1 | 5598 | UTR 3 | XP_016866308.1 |
Set Membership: |
HapMap |