Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAGACAAAATGATGAGGTGGTCT[C/T]GTTTCATCTCCTGTCATCAGTGGTG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610286 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCMT1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LCMT1 - leucine carboxyl methyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032391.1 | Intron | NP_001027563.1 | ||||
NM_016309.2 | Intron | NP_057393.2 | ||||
XM_005255354.3 | Intron | XP_005255411.1 | ||||
XM_011545862.2 | Intron | XP_011544164.1 | ||||
XM_011545864.1 | Intron | XP_011544166.1 |
LCMT1-AS1 - LCMT1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |