Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGCCTCGCAGGAAGGCACCTTGG[A/G]CAGCAGGCTGTGAGGGGCAGTGGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615291 MIM: 616593 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GALT6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
B3GALT6 - beta-1,3-galactosyltransferase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM132A - family with sequence similarity 132 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014980.2 | 669 | Missense Mutation | GCC,GTC | A,V 180 | NP_001014980.1 | |
XM_011541435.2 | 669 | Missense Mutation | GCC,GTC | A,V 180 | XP_011539737.1 | |
XM_011541436.2 | 669 | Missense Mutation | GCC,GTC | A,V 180 | XP_011539738.1 | |
XM_011541437.1 | 669 | Missense Mutation | GCC,GTC | A,V 108 | XP_011539739.1 |
Set Membership: |
HapMap |