Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAAAGCGATGAGGGTAACATGAAA[C/T]TGAAAATCTATGATTTTCACTTTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
28 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607826 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AHCYL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
AHCYL1 - adenosylhomocysteinase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242673.1 | Intron | NP_001229602.1 | ||||
NM_001242674.1 | Intron | NP_001229603.1 | ||||
NM_001242675.1 | Intron | NP_001229604.1 | ||||
NM_001242676.1 | Intron | NP_001229605.1 | ||||
NM_006621.5 | Intron | NP_006612.2 | ||||
XM_011540535.1 | Intron | XP_011538837.1 |
Set Membership: |
HapMap |