Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACTCCCAACTCCCATGAGTCTT[C/T]CCCGCAAGTCCTGCTCCGCCTTCAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
32 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176262 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNA2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KCNA2 - potassium voltage-gated channel subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204269.1 | 11502 | Intron | NP_001191198.1 | |||
NM_004974.3 | 11502 | Intron | NP_004965.1 | |||
XM_011541396.2 | 11502 | UTR 3 | XP_011539698.1 | |||
XM_011541397.2 | 11502 | UTR 3 | XP_011539699.1 | |||
XM_011541398.2 | 11502 | UTR 3 | XP_011539700.1 | |||
XM_011541399.2 | 11502 | UTR 3 | XP_011539701.1 | |||
XM_011541400.2 | 11502 | UTR 3 | XP_011539702.1 | |||
XM_017001213.1 | 11502 | UTR 3 | XP_016856702.1 |
Set Membership: |
HapMap |