Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGGGATCCATTCTGGCTCTTTAGC[A/G]AAAGGTTTGTGAAAGTGACGGATGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176262 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCNA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
KCNA2 - potassium voltage-gated channel subfamily A member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204269.1 | 4102 | Intron | NP_001191198.1 | |||
NM_004974.3 | 4102 | Intron | NP_004965.1 | |||
XM_011541396.2 | 4102 | UTR 3 | XP_011539698.1 | |||
XM_011541397.2 | 4102 | UTR 3 | XP_011539699.1 | |||
XM_011541398.2 | 4102 | UTR 3 | XP_011539700.1 | |||
XM_011541399.2 | 4102 | UTR 3 | XP_011539701.1 | |||
XM_011541400.2 | 4102 | UTR 3 | XP_011539702.1 | |||
XM_017001213.1 | 4102 | UTR 3 | XP_016856702.1 |
Set Membership: |
HapMap |