Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTTTAAAACGTATGAAACAAGC[A/C]AGGGATTATTCTTGCCCCGTGTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
53 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151525 MIM: 615354 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD53 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CD53 - CD53 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000560.3 | Intron | NP_000551.1 | ||||
NM_001040033.1 | Intron | NP_001035122.1 | ||||
NM_001320638.1 | Intron | NP_001307567.1 |
LRIF1 - ligand dependent nuclear receptor interacting factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006945.1 | Intron | NP_001006946.1 | ||||
NM_018372.3 | Intron | NP_060842.3 | ||||
XM_005271029.3 | Intron | XP_005271086.1 | ||||
XM_011541768.1 | Intron | XP_011540070.1 | ||||
XM_017001768.1 | Intron | XP_016857257.1 | ||||
XM_017001769.1 | Intron | XP_016857258.1 |