Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTCTAAGAGTTAATTGTTTATA[C/T]ATAGTGGGACCATCTCAATTTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
23 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601526 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHI3L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CHI3L2 - chitinase 3 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025197.1 | Intron | NP_001020368.1 | ||||
NM_001025199.1 | Intron | NP_001020370.1 | ||||
NM_004000.2 | Intron | NP_003991.2 |
Set Membership: |
HapMap |