Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGGAGTGTCAGAAATAAATGTCAG[A/G]CATCTGCAATCAGCCCTCTTGCTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614234 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PIFO PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PIFO - primary cilia formation | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300831.1 | Intron | NP_001287760.1 | ||||
NM_181643.5 | Intron | NP_857594.2 | ||||
XM_005270472.1 | Intron | XP_005270529.1 | ||||
XM_017000322.1 | Intron | XP_016855811.1 |