Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCCAAACCCAGTTGATGGGACA[G/T]TCATCCTGCCAGGTACTAAGATCAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC61 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CCDC61 - coiled-coil domain containing 61 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001267723.1 | 778 | Intron | NP_001254652.1 | |||
XM_005259191.3 | 778 | Intron | XP_005259248.1 | |||
XM_005259192.4 | 778 | UTR 5 | XP_005259249.1 | |||
XM_011527257.2 | 778 | Intron | XP_011525559.1 |
Set Membership: |
HapMap |