Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGATCCAATCCCCTGCCCCCATGC[C/G]TAAGAAAGAAAAAATGGCCAAGACG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 611088 | ||||||||||||||||||||
Literature Links: |
CCDC65 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC65 - coiled-coil domain containing 65 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286957.1 | 232 | UTR 5 | NP_001273886.1 | |||
NM_033124.4 | 232 | Missense Mutation | CCT,CGT | P,R 2 | NP_149115.2 |