Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTAATTTCAAACTGCAATTCTTAT[C/T]CTTGCAGAAATTGCTAGAATTTATG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602113 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMT2D PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
KMT2D - lysine methyltransferase 2D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RHEBL1 - Ras homolog enriched in brain like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303126.1 | 245 | Intron | NP_001290055.1 | |||
NM_144593.2 | 245 | Intron | NP_653194.1 | |||
XM_017018794.1 | 245 | UTR 5 | XP_016874283.1 | |||
XM_017018795.1 | 245 | UTR 5 | XP_016874284.1 |
Set Membership: |
HapMap |