Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTCGACATTGAAATTGTTTGCA[A/G]CTTTTTGCTATTATAAATAATGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610290 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GALNT12 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GALNT12 - polypeptide N-acetylgalactosaminyltransferase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024642.4 | Intron | NP_078918.3 | ||||
XM_006717287.1 | Intron | XP_006717350.1 | ||||
XM_011519018.2 | Intron | XP_011517320.1 | ||||
XM_011519020.1 | Intron | XP_011517322.1 | ||||
XM_017015133.1 | Intron | XP_016870622.1 |
Set Membership: |
HapMap |