Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAACCAAGGCACTGGGCGCGGTGAG[G/T]CCGTGGGCGCAGAGTCCGAGAATCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616434 | ||||||||||||||||||||
Literature Links: |
IST1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IST1 - IST1, ESCRT-III associated factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270975.1 | Intron | NP_001257904.1 | ||||
NM_001270976.1 | Intron | NP_001257905.1 | ||||
NM_001270977.1 | Intron | NP_001257906.1 | ||||
NM_001270978.1 | Intron | NP_001257907.1 | ||||
NM_001270979.1 | Intron | NP_001257908.1 | ||||
NM_014761.3 | Intron | NP_055576.2 |
ZNF821 - zinc finger protein 821 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |