Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGAGCGGCCTGACCTGGCCCAGA[A/C]GCGCAGCCCTTGGGAAATTCCACAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610336 MIM: 605217 | ||||||||||||||||||||
Literature Links: |
C2CD4C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2CD4C - C2 calcium dependent domain containing 4C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHC2 - SHC adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012435.2 | Intron | NP_036567.2 | ||||
XM_011527893.2 | Intron | XP_011526195.1 | ||||
XM_011527894.2 | Intron | XP_011526196.1 | ||||
XM_011527895.2 | Intron | XP_011526197.1 | ||||
XM_011527896.2 | Intron | XP_011526198.1 | ||||
XM_017026567.1 | Intron | XP_016882056.1 |