Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACGGAACACAGGAGCCCGTGTGCC[A/G]CAGGCCGCAGGCAGTTTCAGGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610336 MIM: 605217 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2CD4C PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2CD4C - C2 calcium dependent domain containing 4C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHC2 - SHC adaptor protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012435.2 | Intron | NP_036567.2 | ||||
XM_011527893.2 | Intron | XP_011526195.1 | ||||
XM_011527894.2 | Intron | XP_011526196.1 | ||||
XM_011527895.2 | Intron | XP_011526197.1 | ||||
XM_011527896.2 | Intron | XP_011526198.1 | ||||
XM_017026567.1 | Intron | XP_016882056.1 |
Set Membership: |
HapMap |