Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGAAGAAATCTGAAAATAAAGC[A/T]ATGGGCTGGCCCTGGGCAGGGGAAT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603413 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TIAL1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TIAL1 - TIA1 cytotoxic granule-associated RNA binding protein-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033925.1 | Intron | NP_001029097.1 | ||||
NM_001323964.1 | Intron | NP_001310893.1 | ||||
NM_001323965.1 | Intron | NP_001310894.1 | ||||
NM_001323967.1 | Intron | NP_001310896.1 | ||||
NM_001323968.1 | Intron | NP_001310897.1 | ||||
NM_001323969.1 | Intron | NP_001310898.1 | ||||
NM_001323970.1 | Intron | NP_001310899.1 | ||||
NM_003252.3 | Intron | NP_003243.1 | ||||
XM_017016605.1 | Intron | XP_016872094.1 | ||||
XM_017016606.1 | Intron | XP_016872095.1 |
Set Membership: |
HapMap |