Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACTTTCATGGAGGTGAGATGTTTC[C/T]TGCATTTTTGGAGTTGTTGCTTTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613128 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
OBFC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
OBFC1 - oligonucleotide/oligosaccharide binding fold containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024928.4 | Intron | NP_079204.2 | ||||
XM_006717976.3 | Intron | XP_006718039.1 | ||||
XM_011540184.2 | Intron | XP_011538486.1 | ||||
XM_017016669.1 | Intron | XP_016872158.1 | ||||
XM_017016670.1 | Intron | XP_016872159.1 |
SH3PXD2A - SH3 and PX domains 2A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |