Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCTCTTCATGTCCAATCTATAGC[C/T]TGCAGTATATTTTTCTTGCTTACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612333 MIM: 601787 MIM: 615204 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR1307 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MIR1307 - microRNA 1307 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDCD11 - programmed cell death 11 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014976.1 | Intron | NP_055791.1 | ||||
XM_005269647.3 | Intron | XP_005269704.1 | ||||
XM_011539538.2 | Intron | XP_011537840.1 | ||||
XM_011539539.2 | Intron | XP_011537841.1 | ||||
XM_011539540.1 | Intron | XP_011537842.1 |
TAF5 - TATA-box binding protein associated factor 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
USMG5 - up-regulated during skeletal muscle growth 5 homolog (mouse) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206426.1 | Intron | NP_001193355.1 | ||||
NM_001206427.1 | Intron | NP_001193356.1 | ||||
NM_032747.3 | Intron | NP_116136.1 |
Set Membership: |
HapMap |