Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCTTGGCTGCTTGGTCGGAGGCGC[C/T]TACACTTACGGCGGCGGGGGCTCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186770 MIM: 612734 | ||||||||||||||||||||
Literature Links: |
TLX1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TLX1 - T-cell leukemia homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195517.1 | 400 | Silent Mutation | GCC,GCT | A,A 54 | NP_001182446.1 | |
NM_005521.3 | 400 | Silent Mutation | GCC,GCT | A,A 54 | NP_005512.1 | |
XM_011539744.2 | 400 | Silent Mutation | GCC,GCT | A,A 54 | XP_011538046.1 | |
XM_011539745.2 | 400 | Silent Mutation | GCC,GCT | A,A 54 | XP_011538047.1 |
TLX1NB - TLX1 neighbor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |